MilliporeSigma
  • Unc-51-like kinase 1/2-mediated endocytic processes regulate filopodia extension and branching of sensory axons.
Unc-51-like kinase 1/2-mediated endocytic processes regulate filopodia extension and branching of sensory axons.
Proceedings of the National Academy of Sciences of the United States of America (2007-03-29)
Xiang Zhou, J Ramesh Babu, Susana da Silva, Qing Shu, Isabella A Graef, Tim Oliver, Toshifumi Tomoda, Tomomi Tani, Marie W Wooten, Fan Wang
ABSTRACT
The molecular mechanism and significance of endocytic processes involved in directional axon elongation are not well understood. The Unc-51 family of serine/threonine kinases was shown to be important for axon growth and was also linked to endocytosis, providing an entry point to study this problem. We found that mouse Unc-51-like kinase 1/2 (Ulk1/2) proteins are localized to vesicular structures in growth cones of mouse spinal sensory neurons. RNAi-mediated knockdown of Ulk1 and/or Ulk2 resulted in impaired endocytosis of nerve growth factor (NGF), excessive axon arborization, and severely stunted axon elongation. The evidence also indicates that Ulk1/2 mediates a non-clathrin-coated endocytosis in sensory growth cones. Interestingly, NGF can induce the interaction of Ulk1 with TrkA receptor complexes through promoting K63-polyubiquitination of Ulk1 and binding of Ulk1 to the scaffolding protein p62. These results and additional studies suggest that Ulk1/2 proteins regulate filopodia extension and neurite branching during sensory axon outgrowth, probably through regulating TrkA receptor trafficking and signaling.
Sigma-Aldrich
Anti-ULK4 antibody produced in rabbit, affinity isolated antibody