MilliporeSigma
  • Synthesis and antiviral activity of novel trans-2,2-dimethyl cyclopropyl nucleosides.
Synthesis and antiviral activity of novel trans-2,2-dimethyl cyclopropyl nucleosides.
Archives of pharmacal research (2003-12-10)
Min Chul Kook, Bo Gil Choi
ABSTRACT
Novel trans-2,2-dimethylcyclopropyl nucleosides were synthesized as potential antiviral agents. The key intermediate, 3, was synthesized via five steps from ethyl chrysanthemate and condensed with purine bases using the Mitsunobu reaction to give six cyclopropyl nucleosides. These synthesized nucleosides did not show any significant antiviral activity against HSV-1, HSV-2, EMCV, Cox B3, or VSV, at concentrations up to 100 microM.