MilliporeSigma
  • trans-1-[(2-Phenylcyclopropyl)methyl]-4-arylpiperazines: mixed dopamine D(2)/D(4) receptor antagonists as potential antipsychotic agents.
trans-1-[(2-Phenylcyclopropyl)methyl]-4-arylpiperazines: mixed dopamine D(2)/D(4) receptor antagonists as potential antipsychotic agents.
Journal of medicinal chemistry (2000-10-29)
X Zhang, K Hodgetts, S Rachwal, H Zhao, J W Wasley, K Craven, R Brodbeck, A Kieltyka, D Hoffman, M D Bacolod, B Girard, J Tran, A Thurkauf
ABSTRACT
The dopaminergic receptor profile of a series of trans-1-[(2-phenylcyclopropyl)methyl]-4-arylpiperazines was examined. Aromatic substitution patterns were varied with the goal of identifying a compound having affinities for the D(2) and D(4) receptors in a ratio similar to that observed for the atypical neuroleptic clozapine. The compounds (1S, 2S)-trans-1-[(2-phenylcyclopropyl)methyl]-4-(2, 4-dichlorophenyl)piperazine (5m) and (1S, 2S)-trans-1-[(2-phenylcyclopropyl)methyl]-4-(2, 4-dimethylphenyl)piperazine (5t) were selected for functional antagonists at D(2) and D(4) receptors and had a D(2)/D(4) ratio approximating that of clozapine; they proved inactive in behavioral tests of antipsychotic activity.