MilliporeSigma
  • Metabolism of the psychotomimetic tryptamine derivative 5-methoxy-N,N-diisopropyltryptamine in humans: identification and quantification of its urinary metabolites.
Metabolism of the psychotomimetic tryptamine derivative 5-methoxy-N,N-diisopropyltryptamine in humans: identification and quantification of its urinary metabolites.
Drug metabolism and disposition: the biological fate of chemicals (2005-11-11)
Tooru Kamata, Munehiro Katagi, Hiroe T Kamata, Akihiro Miki, Noriaki Shima, Kei Zaitsu, Mayumi Nishikawa, Einosuke Tanaka, Katsuya Honda, Hitoshi Tsuchihashi
ABSTRACT
The urinary metabolites of 5-methoxy-N,N-diisopropyltryptamine (5-MeO-DIPT) in humans have been investigated by analyzing urine specimens from its users. For the unequivocal identification and accurate quantification of its major metabolites, careful analyses were conducted by gas chromatography/mass spectrometry, liquid chromatography/mass spectrometry, and liquid chromatography-tandem mass spectrometry, using authentic standards of each metabolite synthesized. Three major metabolic pathways were revealed as follows: 1) side chain degradation by O-demethylation to form 5-hydroxy-N,N-diisopropyltryptamine (5-OH-DIPT), which would be partly conjugated to its sulfate and glucuronide; 2) direct hydroxylation on position 6 of the aromatic ring of 5-MeO-DIPT, and/or methylation of the hydroxyl group on position 5 after hydroxylation on position 6 of the aromatic ring of 5-OH-DIPT, to produce 6-hydroxy-5-methoxy-N,N-diisopropyltryptamine (6-OH-5-MeO-DIPT), followed by conjugation to its sulfate and glucuronide; and 3) side chain degradation by N-deisopropylation, to the corresponding secondary amine 5-methoxy-N-isopropyltryptamine (5-MeO-NIPT). Of these metabolites, which retain structural characteristics of the parent drug, 5-OH-DIPT and 6-OH-5-MeO-DIPT were found to be more abundant than 5-MeO-NIPT. Although the parent drug 5-MeO-DIPT was detectable even 35 h after dosing, no trace of its N-oxide was detected in any of the specimens examined.
Sigma-Aldrich
5-Methyltryptamine hydrochloride, 98%