MilliporeSigma
  • Lithiation directed by amino alkoxides. Application to the synthesis of new disubstituted dihydrodipyridopyrazines.
Lithiation directed by amino alkoxides. Application to the synthesis of new disubstituted dihydrodipyridopyrazines.
Organic letters (2006-09-08)
Irina-Claudia Grig-Alexa, Adriana-Luminita Fînaru, Paul Caubère, Gérald Guillaumet
ABSTRACT
The synthesis of new 4,6-disubstituted dihydrodipyridopyrazines starting from corresponding carboxaldehydes via lithiation directed by alpha-amino alkoxides is described. The N,N,N'-trimethylethylenediamine was used as amine component for in situ formation of the alpha-amino alkoxides. After optimization, this reaction allowed easy access to new interesting starting materials for further applications by palladium-catalyzed reactions.
Sigma-Aldrich
N,N,N′-Trimethylethylenediamine, 97%