MilliporeSigma
  • Molten-globule state of carbonic anhydrase binds to the chaperone-like alpha-crystallin.
Molten-globule state of carbonic anhydrase binds to the chaperone-like alpha-crystallin.
The Journal of biological chemistry (1996-11-01)
K Rajaraman, B Raman, C M Rao
ABSTRACT
alpha-Crystallin, a multimeric protein, exhibits chaperone-like activity in preventing aggregation of several proteins. We have studied the chaperone-like activity of alpha-crystallin toward heat-induced aggregation of bovine and human carbonic anhydrase. Human carbonic anhydrase aggregates at 60 degrees C, while bovine carbonic anhydrase does not aggregate significantly at this temperature. Removal of the enzyme-bound metal ion, Zn2+, by EDTA modulates the aggregation behavior of bovine carbonic anhydrase. Fluorescence and circular dichroism studies show that removal of the metal ion from the bovine carbonic anhydrase by a chelator such as EDTA enhances the propensity of the enzyme to adopt the molten-globule state. alpha-Crystallin binds to this state of the enzyme and prevents aggregation. Fluorescence and circular dichroism studies on the alpha-crystallin-enzyme complexes show that the enzymes in the complex are in the molten-globule state. These results are of relevance to the interaction of chaperones with the partially unfolded states of target proteins.
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein