MilliporeSigma
  • TNFSF10 (TRAIL), a p53 target gene that mediates p53-dependent cell death.
TNFSF10 (TRAIL), a p53 target gene that mediates p53-dependent cell death.
Cancer biology & therapy (2008-12-25)
Kageaki Kuribayashi, Gabriel Krigsfeld, Wenge Wang, Jing Xu, Patrick A Mayes, David T Dicker, Gen Sheng Wu, Wafik S El-Deiry
ABSTRACT
We have identified TNFSF10 (TRAIL) as a p53-transcriptional target gene. There are two p53 DNA-binding sites in the human TNFSF10 promoter region, at 346 and 625 bp upstream of the transcription start site. A human p53-expressing adenovirus (Ad-p53) induced TRAIL mRNA and protein expression in HCT116 p53-/- human colon cancer cells. A human TRAIL-promoter reporter assay showed increased luciferase activity with the promoter vector that contains two p53 DNA-binding motifs,following Ad-p53 infection, compared to the control adenovirus infection. Using HCT116 cells, gene silencing of TNFSF10 by siRNA suppressed caspase 3 and 7 activity, even after treatment with the DNA-damaging chemotherapeutic agent adriamycin. TRAIL protein expression was elevated in adriamycin-treated breast cancer cells. In vivo, TRAIL expression was induced in mouse natural killer cells at 24 hours after systemic treatment with 5-Fluorouracil. p53-dependent TRAIL induction in natural killer cells after chemotherapy exposure provides a link between the tumor suppressor p53 and the host immune response during cancer therapy as well as a paracrine-mediated cell-extrinsic death response. Our findings provide new mechanistic insights into the signaling of p53-dependent cell death and tumor suppression, including the involvement of the host immune system and natural killer cells in vivo in the anti-tumor efficacy of chemotherapy.