MilliporeSigma
  • Incorporation of 1-deoxy-D-xylulose into isoprene and phytol by higher plants and algae.
Incorporation of 1-deoxy-D-xylulose into isoprene and phytol by higher plants and algae.
FEBS letters (1997-09-26)
J Schwender, J Zeidler, R Gröner, C Müller, M Focke, S Braun, F W Lichtenthaler, H K Lichtenthaler
ABSTRACT
In further substantiating the novel mevalonate-independent pathway for isoprenoid biosynthesis, which generates isopentenyl diphosphate (IPP) via 1-deoxy-D-xylulose-5-phosphate, labeling experiments with 1-[2H(1)]deoxy-D-xylulose were performed with various higher plants and algae: efficient incorporation was observed into isoprene emitted by Populus, Chelidonium, and Salix, into the phytol moiety of chlorophylls in a red alga (Cyanidium), in two green algae (Scenedesmus, Chlamydomonas), and a higher plant (Lemna). By contrast, 13C-mevalonate applied was incorporated into isoprene and phytol to a much lower extent or not at all. This demonstrates that this '1-deoxy-D-xylulose-5-phosphate pathway' for biosynthesis of plastidic isoprenoids is widely distributed in photosynthetic organisms.
Sigma-Aldrich
1-Deoxy-D-xylulose-5-phosphate sodium salt, ≥99.0% (TLC)