MilliporeSigma
  • Growth suppression and induction of heat-shock protein-70 by 9-cis beta-carotene in cervical dysplasia-derived cells.
Growth suppression and induction of heat-shock protein-70 by 9-cis beta-carotene in cervical dysplasia-derived cells.
Life sciences (1997-01-01)
T Toba, Y Shidoji, J Fujii, H Moriwaki, Y Muto, T Suzuki, N Ohishi, K Yagi
ABSTRACT
The aim of the present study was to determine to what extent 9-cis beta-carotene, one of the most abundant naturally-occurring cis-isomers of beta-carotene, can inhibit the growth of cervical dysplasia-derived cells in comparison with all-trans beta-carotene. We found that 9-cis beta-carotene was dose-dependently more effective than all-trans beta-carotene. Both carotenes induced the intracellular accumulation of heat-shock protein-70 (HSP70), and the treated cells showed morphological changes indicative of apoptosis. The results of the present study strongly suggest that the induction of HSP70 by beta-carotene might be involved in beta-carotene-mediated suppression of the cell growth through apoptosis.
Sigma-Aldrich
(9Z)-β-Carotene, ≥90.0% (HPLC)