MilliporeSigma
  • A functional carbohydrate chip platform for analysis of carbohydrate-protein interaction.
A functional carbohydrate chip platform for analysis of carbohydrate-protein interaction.
Nanotechnology (2010-05-01)
Jeong Hyun Seo, Chang Sup Kim, Byeong Hee Hwang, Hyung Joon Cha
ABSTRACT
A carbohydrate chip based on glass or other transparent surfaces has been suggested as a potential tool for high-throughput analysis of carbohydrate-protein interactions. Here we proposed a facile, efficient, and cost-effective method whereby diverse carbohydrate types are modified in a single step and directly immobilized onto a glass surface, with retention of functional orientation. We modified various types of carbohydrates by reductive amination, in which reducing sugar groups were coupled with 4-(2-aminoethyl)aniline, which has di-amine groups at both ends. The modified carbohydrates were covalently attached to an amino-reactive NHS-activated glass surface by formation of stable amide bonds. This proposed method was applied for efficient construction of a carbohydrate microarray to analyze carbohydrate-protein interactions. The carbohydrate chip prepared using our method can be successfully used in diverse biomimetic studies of carbohydrates, including carbohydrate-biomolecule interactions, and carbohydrate sensor chip or microarray development for diagnosis and screening.
Sigma-Aldrich
4-(2-Aminoethyl)aniline, 97%