MilliporeSigma
Dietary citric acid enhances absorption of aluminum in antacids.
Clinical chemistry (1986-03-01)
P Slanina, W Frech, L G Ekström, L Lööf, S Slorach, A Cedergren
ABSTRACT
Ten healthy men ingested, twice daily between meals, during each of the seven-day experimental periods: (a) citric acid (as lemon juice), (b) Al(OH)3, or (c) Al(OH)3 + citric acid. Whole blood sampled after each dietary period was analyzed electrothermally after digestion with nitric acid. Moderate, but significant, increases in mean Al concentrations as compared with pretreatment values [5 (SD 3) micrograms of Al per liter] were seen after ingestion of either citric acid or Al(OH)3: 9 (SD 4) and 12 (SD 3) micrograms/L, respectively. Ingestion of both Al(OH)3 and citric acid resulted in a more pronounced, highly significant (p less than 0.001) increase in Al concentrations, to 23 (SD 2) micrograms Al/L, probably owing to formation and absorption of Al-citrate complexes.
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)