MilliporeSigma
  • Preparation, characterization and photocatalytic behavior of WO3-fullerene/TiO2 catalysts under visible light.
Preparation, characterization and photocatalytic behavior of WO3-fullerene/TiO2 catalysts under visible light.
Nanoscale research letters (2011-07-22)
Ze-Da Meng, Lei Zhu, Jong-Geun Choi, Chong-Yeon Park, Won-Chun Oh
ABSTRACT
WO3-treated fullerene/TiO2 composites (WO3-fullerene/TiO2) were prepared using a sol-gel method. The composite obtained was characterized by BET surface area measurements, X-ray diffraction, scanning electron microscopy, energy dispersive X-ray analysis, transmission electron microscopy, and UV-vis analysis. A methyl orange (MO) solution under visible light irradiation was used to determine the photocatalytic activity. Excellent photocatalytic degradation of a MO solution was observed using the WO3-fullerene, fullerene-TiO2, and WO3-fullerene/TiO2 composites under visible light. An increase in photocatalytic activity was observed, and WO3-fullerene/TiO2 has the best photocatalytic activity; it may attribute to the increase of the photo-absorption effect by the fullerene and the cooperative effect of the WO3.
Sigma-Aldrich
Ammonium metatungstate hydrate, ≥85% WO3 basis (gravimetric)
Sigma-Aldrich
Ammonium metatungstate hydrate, ≥66.5% (W)
Sigma-Aldrich
Ammonium metatungstate hydrate, 99.99% trace metals basis