MilliporeSigma
  • (E)-3-tert-butyl-4-(4-chlorobenzyl)-N-(4-chlorobenzylidene)-1-phenyl-1H-pyrazol-5-amine: sheets built from π-stacked hydrogen-bonded dimers.
(E)-3-tert-butyl-4-(4-chlorobenzyl)-N-(4-chlorobenzylidene)-1-phenyl-1H-pyrazol-5-amine: sheets built from π-stacked hydrogen-bonded dimers.
Acta crystallographica. Section C, Crystal structure communications (2013-09-06)
Jairo Quiroga, Dayana Pantoja, Justo Cobo, Christopher Glidewell
ABSTRACT
The title compound, C27H25Cl2N3, is an unexpected but high-yield product from the microwave-mediated reaction between 3-tert-butyl-N-4-chlorobenzyl-1-phenyl-1H-pyrazol-5-amine and 4-chlorobenzaldehyde. Inversion-related pairs of molecules are linked by C-H···π(arene) hydrogen bonds to form cyclic centrosymmetric dimers, and dimers of this type are linked into sheets by two independent π-π stacking interactions.
Sigma-Aldrich
4-Chlorobenzaldehyde, 97%