MilliporeSigma
  • Albumin stimulates interleukin-8 expression in proximal tubular epithelial cells in vitro and in vivo.
Albumin stimulates interleukin-8 expression in proximal tubular epithelial cells in vitro and in vivo.
The Journal of clinical investigation (2003-02-18)
Sydney Tang, Joseph C K Leung, Katsushige Abe, Kwok Wah Chan, Loretta Y Y Chan, Tak Mao Chan, Kar Neng Lai
ABSTRACT
Renal tubulointerstitial injury is characterized by inflammatory cell infiltrate; however, the stimuli for leukocyte recruitment are not fully understood. IL-8 is a potent chemokine produced by proximal tubular epithelial cells (PTECs). Whether nephrotic proteins stimulate tubular IL-8 expression remains unknown. Acute exposure of human PTECs to albumin induced IL-8 gene and protein expression time- and dose-dependently. Apical albumin predominantly stimulated basolateral IL-8 secretion. Electrophoretic mobility shift assay demonstrated nuclear translocation of NF-kappaB, and the p65/p50 subunits were activated. NF-kappaB activation and IL-8 secretion were attenuated by the NF-kappaB inhibitors pyrrolidine dithiocarbamate and cell-permeable peptide. Albumin upregulated intracellular reactive oxygen species (ROS) generation, while exogenous H2O2 stimulated NF-kappaB translocation and IL-8 secretion. Albumin-induced ROS generation, NF-kappaB activation, and IL-8 secretion were endocytosis- and PKC-dependent as these downstream events were abrogated by the PI3K inhibitors LY294002 and wortmannin, and the PKC inhibitors GF109203X and staurosporin, respectively. In vivo, IL-8 mRNA expression was localized by in situ hybridization to the proximal tubules in nephrotic kidney tissues. The intensity of IL-8 immunostaining was higher in nephrotic than non-nephrotic subjects. In conclusion, albumin is a strong stimulus for tubular IL-8 expression, which occurs via NF-kappaB-dependent pathways through PKC activation and ROS generation.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization