MilliporeSigma
  • Percutaneous penetration of benzene and benzene contained in solvents used in the rubber industry.
Percutaneous penetration of benzene and benzene contained in solvents used in the rubber industry.
Archives of environmental health (1981-09-01)
H I Maibach, D M Anjo
ABSTRACT
Penetration of benzene through the skin of the rhesus monkey was determined using 14C-benzene, and quantitating the labelled metabolites in urine. The modes of application and amounts of benzene that penetrated the skin (indicated in parentheses) are as follows: (1) a single, direct cutaneous application of liquid benzene (0.172 +/- 0.139%); (2) a single application of benzene-containing [0.36%] solvent (0.0805 +/- 0.03060%); (3) multiple washes with full-strength benzene (0.848 +/- 0.0806%); (4) multiple washes with the benzene-containing [0.35%] solvent (0.431 +/- 0.258%); (5) removal of the stratum corneum followed by application of full-strength benzene (0.909 +/- 0.627%); and (6) application of benzene to the palmar surface (0.651 +/- 0.482%). Until more complete human data becomes available, benzene penetration in the monkey may be used to estimate penetration in man, both for industrial hygiene purposes and general toxicological use.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard