MilliporeSigma
  • Biochemical and biomedical applications of capillary column GLC using SIM.
Biochemical and biomedical applications of capillary column GLC using SIM.
Journal of chromatographic science (1983-03-01)
W J VandenHeuvel, J R Carlin, R W Walker
ABSTRACT
This report focuses on recent applications of capillary column GLC to the analysis of drugs and metabolites, other xenobiotics, natural products, and environmental contaminants in samples of biological origin. The increasing use of selected ion monitoring, combined with stable isotope methods as a means of GLC detection and quantification, is emphasized. Specific topics covered include the use of capillary column GLC for 1) determining the pharmacokinetics of timolol (BLOCADREN) in human volunteers by simultaneous oral and intravenous administration of this beta-blocker and its 13C3-labeled analog; 2) measuring residue levels of the forced molting agent, xylonidine, in yolk and albumen of chicken eggs; 3) monitoring animal plasma and tissues for the depletion of 5-hydroxy-1,3-dioxane and 4-hydroxymethyl-1,3-dioxolane (isomeric components of glycerol formal, an animal drug formulation component); 4) detecting 13C, 15N-methyltryptamine (a biosynthesis product from 13C, 15N-tryptamine) in human urine; and 5) measuring estradiol levels in biological samples.
Sigma-Aldrich
N-ω-Methyltryptamine, 99%