MilliporeSigma
The early history of estriol.
Journal of steroid biochemistry (1984-04-01)
E Diczfalusy
ABSTRACT
The early history of estriol is reviewed with special emphasis on its isolation, identification, quantitation in tissues and body fluids and its unique biogenesis in the human feto-placental unit. The relationship to epimeric estriols and acidic estrogens and the pharmacokinetic and pharmacodynamic properties which led to a widespread therapeutic use of this compound are also discussed.
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
Estriol, ≥97%
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®