MilliporeSigma
  • Amniotic fluid lecithin/sphingomyelin ratio, palmitic acid, palmitic acid/stearic acid ratio, total cortisol, creatinine, and percentage of lipid-positive cells in assessment of fetal maturity and fetal pulmonary maturity: a comparison.
Amniotic fluid lecithin/sphingomyelin ratio, palmitic acid, palmitic acid/stearic acid ratio, total cortisol, creatinine, and percentage of lipid-positive cells in assessment of fetal maturity and fetal pulmonary maturity: a comparison.
American journal of obstetrics and gynecology (1979-02-01)
T A Doran, J A Ford, L C Allen, P Y Wong, R J Benzie
PMID433991
ABSTRACT
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples were taken within 72 hours of delivery. The best tests for fetal maturity (37 weeks) with differential percentages were L/S ratio, palmitic acid, and P/S ratio. In the assessment of fetal pulmonary maturity, we studied an additional 174 samples in which only L/S ratio, creatinine, and lipid-positive cells were analyzed. All tests showed a high predictive value of an immature (positive) result was much less for all six parameters; the three best tests were total cortisol (33%), lipid-positive cells (26%) and L/S ratio (14%).
Product Description
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Stearic acid, European Pharmacopoeia (EP) Reference Standard