MilliporeSigma
  • Octanoic acidemia and octanoylcarnitine excretion with dicarboxylic aciduria due to defective oxidation of medium-chain fatty acids.
Octanoic acidemia and octanoylcarnitine excretion with dicarboxylic aciduria due to defective oxidation of medium-chain fatty acids.
The Journal of pediatrics (1985-09-01)
M Duran, G Mitchell, J B de Klerk, J P de Jager, M Hofkamp, L Bruinvis, D Ketting, J M Saudubray, S K Wadman
ABSTRACT
Five patients aged 7 to 21 months are described who developed attacks of coma after a short prodromal illness with diarrhea or vomiting or both. Four had concomitant hypoglycemia, and all had hypoketonemia, with excessive urinary excretion of medium-chain dicarboxylic acids, medium-chain (omega-1)-hydroxyacids, suberylglycine, hexanoylglycine, and octanoylcarnitine. All patients accumulated octanoic acid, decanoic acid, and cis-4-decenoic acid in plasma. Fibroblasts from three patients showed a decreased rate of octanoate oxidation (10%, 12%, and 29% of control values, respectively). These findings suggest a deficiency of medium-chain acyl-CoA dehydrogenase, most probably an autosomal recessive inherited metabolic disorder. Two of the patients died during an acute attack, and a third had severe neurologic sequelae; the two remaining patients recovered. Plasma free carnitine levels were low, but total carnitine was normal. The three surviving patients underwent a fasting test, which did not lead to hypoglycemia, although hypoketonemia, dicarboxylic aciduria, and excessive mobilization of fatty acids did occur. The surviving patients were maintained on frequent carbohydrate-enriched meals.
Product Description
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥98.0%