MilliporeSigma
Liquid/air partition coefficients of four terpenes.
British journal of industrial medicine (1990-01-01)
A Falk, E Gullstrand, A Löf, E Wigaeus-Hjelm
ABSTRACT
The liquid/air partition coefficients of four common terpenes, alpha-pinene, beta-pinene, 3-carene, and limonene, have been determined in vitro using head space technique. The liquids used were water, human blood, and olive oil. alpha-Pinene, beta-pinene, and 3-carene were practically insoluble in water and limonene was slightly soluble; all were readily dissolved in olive oil. The oil/air partition coefficients ranged from 2900 to 5700 in the order alpha-pinene, beta-pinene, 3-carene, and limonene. The blood/air partition coefficients ranged from 15 to 42 in the same order as for oil/air.
Sigma-Aldrich
(S)-(−)-Limonene, 96%
Sigma-Aldrich
(S)-(−)-Limonene, ≥95%, FG
Supelco
(S)-(−)-Limonene, analytical standard