MilliporeSigma
  • Apigenin-induced-apoptosis is mediated by the activation of PKCdelta and caspases in leukemia cells.
Apigenin-induced-apoptosis is mediated by the activation of PKCdelta and caspases in leukemia cells.
Biochemical pharmacology (2006-07-18)
Melissa A Vargo, Oliver H Voss, Frantisek Poustka, Arturo J Cardounel, Erich Grotewold, Andrea I Doseff
ABSTRACT
Apigenin, a flavone abundantly found in fruits and vegetables, exhibits antiproliferative, anti-inflammatory, and antimetastatic activities through poorly defined mechanisms. In the present study, the treatment of different cell lines with apigenin resulted in selective antiproliferative and apoptotic effect in monocytic and lymphocytic leukemias. Apigenin-induced-apoptosis was mediated by the activation of caspase-9 and caspase-3. Apigenin was found intracellularly and localized to the mitochondria. Treatment of monocytic cells with apigenin was accompanied by an increase in reactive oxygen species (ROS) and phosphorylation of the MAPKs, p38 and ERK. However, the inhibition of ROS, p38 or ERK failed to block apoptosis, suggesting that these cellular responses induced by apigenin are not essential for the induction of apoptosis. In addition, apigenin induced the activation of PKCdelta. Pharmacological inhibition of PKCdelta, the expression of dominant-negative PKCdelta and silencing of PKCdelta in leukemia cells showed that apigenin-induced-apoptosis requires PKCdelta activity. Together, these results indicate that this flavonoid provides selective activity to promote caspase-dependent-apoptosis of leukemia cells and uncover an essential role of PKCdelta during the induction of apoptosis by apigenin.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Supelco
Apigenin, analytical standard