MilliporeSigma
Metabolomics in human nutrition: opportunities and challenges.
The American journal of clinical nutrition (2005-09-13)
Michael J Gibney, Marianne Walsh, Lorraine Brennan, Helen M Roche, Bruce German, Ben van Ommen
ABSTRACT
Metabolomics has been widely adopted in pharmacology and toxicology but is relatively new in human nutrition. The ultimate goal, to understand the effects of exogenous compounds on human metabolic regulation, is similar in all 3 fields. However, the application of metabolomics to nutritional research will be met with unique challenges. Little is known of the extent to which changes in the nutrient content of the human diet elicit changes in metabolic profiles. Moreover, the metabolomic signal from nutrients absorbed from the diet must compete with the myriad of nonnutrient signals that are absorbed, metabolized, and secreted in both urine and saliva. The large-bowel microflora also produces significant metabolic signals that can contribute to and alter the metabolome of biofluids in human nutrition. Notwithstanding these possible confounding effects, every reason exists to be optimistic about the potential of metabolomics for the assessment of various biofluids in nutrition research. This potential lies both in metabolic profiling through the use of pattern-recognition statistics on assigned and unassigned metabolite signals and in the collection of comprehensive data sets of identified metabolites; both objectives have the potential to distinguish between different dietary treatments, which would not have been targeted with conventional techniques. The latter objective sets out a well-recognized challenge to modern biology: the development of libraries of small molecules to aid in metabolite identification. The purpose of the present review was to highlight some early challenges that need to be addressed if metabolomics is to realize its great potential in human nutrition.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
17α-Hydroxyprogesterone, ≥95%
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
17α-Hydroxyprogesterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Estradiol, meets USP testing specifications