MilliporeSigma
  • Comparison of the levels of copper, zinc and iron in cervical mucus and in blood serum of women of childbearing age.
Comparison of the levels of copper, zinc and iron in cervical mucus and in blood serum of women of childbearing age.
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina (2005-09-09)
Andrzej Kaczmarek, Dorota Robak-Chołubek, Ireneusz Sowa, Grzegorz Jakiel
ABSTRACT
The objective of this research study was a preliminary comparison of some chosen trace elements concentrations in cervical mucus and blood serum of women of childbearing age. The levels of iron (Fe), copper (Cu) and zinc (Zn) were determined in cervical mucus and serum of 10 patients. The obtained results do not point to any association between the levels of the examined trace elements in cervical mucus or any correlation between their contents in mucus and blood serum. As no correlation was found between the blood serum and cervical mucus, as far as trace elements are concerned, one might suggest that the uptake of trace elements by the mucus has an active form.
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%