Merck
  • Structural determination of novel lacto-N-decaose and its monofucosylated analogue from human milk by electrospray tandem mass spectrometry and 1H NMR spectroscopy.
Structural determination of novel lacto-N-decaose and its monofucosylated analogue from human milk by electrospray tandem mass spectrometry and 1H NMR spectroscopy.
Archives of biochemistry and biophysics (2005-01-05)
Wengang Chai, Vladimir E Piskarev, Yibing Zhang, Alexander M Lawson, Heide Kogelberg
ABSTRACT
We have isolated and characterised two neutral oligosaccharides, one nonfucosylated and the other monofucosylated, from human milk that are based on the doubly branched lacto-N-decaose core. Their structures have been determined by a combined use of electrospray tandem mass spectrometry (ES-MS/MS) and NMR spectroscopy. The sequences of the three branches resulted from the double-branching, including the identity and location of the blood-group-related Lewis determinant and partial linkages, were elucidated by the unique method of high sensitivity negative-ion ES-MS/MS analysis. Their full structure assignment was completed by methylation analysis and 1H NMR. The monofucosylated lacto-N-decaose, Galbeta1-4(Fucalpha1-3)GlcNAcbeta1-6(Galbeta1-3GlcNAcbeta1-3)Galbeta1-4GlcNAcbeta1-6(Galbeta1-3GlcNAcbeta1-3)Galbeta1-4Glc is a novel sequence, whereas the nonfucosylated lacto-N-decaose, Galbeta1-4GlcNAcbeta1-6(Galbeta1-3GlcNAcbeta1-3)Galbeta1-4GlcNAcbeta1-6(Galbeta1-3GlcNAcbeta1-3)Galbeta1-4Glc, has not been isolated and identified as an individual oligosaccharide.