MilliporeSigma
  • Glycerophosphocholine is elevated in cerebrospinal fluid of Alzheimer patients.
Glycerophosphocholine is elevated in cerebrospinal fluid of Alzheimer patients.
Neurobiology of aging (2004-10-07)
Anna Walter, Ulrike Korth, Michael Hilgert, Joachim Hartmann, Oksana Weichel, Marion Hilgert, Klaus Fassbender, Andrea Schmitt, Jochen Klein
ABSTRACT
Experimental and clinical studies give evidence for breakdown of membrane phospholipids during neurodegeneration. In the present study, we measured the levels of glycerophosphocholine (GPCh), phosphocholine (PCh), and choline, that is, water-soluble metabolites of phosphatidylcholine (PtdCho), in human cerebrospinal fluid (CSF). Among 30 cognitively normal patients the average CSF levels of GPCh, phosphocholine and choline were 3.64, 1.28, and 1.93 microM, respectively; metabolite levels did not change with increasing age. When compared with age-matched controls, patients with Alzheimer's disease had elevated levels of all choline metabolites: GPCh was significantly increased by 76% (P<0.01), phosphocholine by 52% (P<0.05), and free choline (Ch) by 39%. Six patients with vascular dementia had lower choline and elevated phosphocholine levels, when compared to controls, but normal levels of GPCh. These data demonstrate that Alzheimer's disease is accompanied by an increased PtdCho hydrolysis in the brain. PtdCho breakdown seems to be mediated by phospholipase A2 and leads to significantly elevated levels of GPCh in CSF.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination