MilliporeSigma
Wilson disease.
Virchows Archiv : an international journal of pathology (2004-06-19)
Cord Langner, Helmut Denk
ABSTRACT
Wilson disease (WD) is an autosomal recessive disorder of copper metabolism. Since daily copper intake exceeds the body's requirements, effective means of excreting excess copper are essential. These are accomplished by ATP7B, a new member of the cation-transporting p-type ATPase family, which is mainly expressed in the liver and mediates both copper secretion into plasma (coupled with ceruloplasmin synthesis) and its excretion into bile. Thus far, more than 200 mutations of the WD gene have been detected, causing impairment of ATP7B function and, ultimately, copper accumulation. Excess copper, however, induces free-radical reactions and lipid peroxidation. Resultant liver damage leads to steatosis, inflammation, cirrhosis, and, occasionally, fulminant liver failure. The diagnosis of WD is commonly made on the basis of typical clinical and laboratory findings, including low serum ceruloplasmin, increased urinary copper excretion, and increased hepatic copper content. Since liver morphology is non-specific, and copper histochemistry may lead to both false-negative and false-positive results, the pathologist usually only suspects the disease or assists in its confirmation. Although the value of molecular genetic testing is limited due to the high number of possible gene mutations, polymerase chain reaction may be useful for the evaluation of family members of homozygous index patients.
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, 99.999% trace metals basis
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Copper, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis