MilliporeSigma
  • Nongenomic effects of 17beta-estradiol in human platelets: potentiation of thrombin-induced aggregation through estrogen receptor beta and Src kinase.
Nongenomic effects of 17beta-estradiol in human platelets: potentiation of thrombin-induced aggregation through estrogen receptor beta and Src kinase.
Blood (2004-06-17)
Laura Moro, Stefania Reineri, Daniela Piranda, Daniela Pietrapiana, Paolo Lova, Alessandra Bertoni, Andrea Graziani, Paola Defilippi, Ilaria Canobbio, Mauro Torti, Fabiola Sinigaglia
ABSTRACT
The impact of estrogens on the cardiovascular system and their ability to regulate platelet function are matters of controversy. The recent finding that estrogen receptors are expressed in human platelets renders these cells an excellent model for studying the nongenomic effects of these hormones. In this work, we investigated 17beta-estradiol-dependent signaling in platelets from adult healthy men. 17beta-estradiol caused the rapid phosphorylation of the tyrosine kinases Src and Pyk2 and the formation of a signaling complex, which included Src, Pyk2, and the phosphatidylinositol 3-kinase. Both these events were dependent on estrogen receptor beta engagement. We found that estrogen receptor beta was membrane-associated in platelets. On treatment with 17beta-estradiol, Src and Pyk2 activation occurred in the membrane fraction but not in the cytosol. In contrast, no significant activation of phosphatidylinositol 3-kinase was detected in estrogen-treated platelets. 17beta-estradiol did not induce any platelet response directly, but it strongly potentiated the activation of integrin alpha(IIb)beta3 and the platelet aggregation induced by subthreshold concentrations of thrombin. These effects were dependent on estrogen receptor beta recruitment and were associated with a strong synergistic effect with thrombin on Src activation. Taken together, these results indicate that 17beta-estradiol can modulate platelet function by exercising a proaggregating role.
Product Description
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard