MilliporeSigma
  • Protection from obesity and insulin resistance in mice overexpressing human apolipoprotein C1.
Protection from obesity and insulin resistance in mice overexpressing human apolipoprotein C1.
Diabetes (2001-11-28)
M C Jong, P J Voshol, M Muurling, V E Dahlmans, J A Romijn, H Pijl, L M Havekes
ABSTRACT
Apolipoprotein (APO) C1 is a 6.6-kDa protein present in plasma and associated with lipoproteins. Using hyperinsulinemic-euglycemic clamp tests, we previously found that in APOC1 transgenic mice, the whole-body insulin-mediated glucose uptake is increased concomitant with a decreased fatty acid uptake. These latter results are confirmed in the present study, showing that APOC1 transgenic mice exhibit a 50% reduction in the uptake of the fatty acid analog 15-(p-iodophenyl)-3-(R,S)-methyl pentadecanoic acid in white adipose tissue stores. We next investigated whether APOC1 overexpression can modulate the initiation and/or development of obesity and insulin resistance. When crossbred on the genetically obese ob/ob background, APOC1 transgenic mice were fully protected from the development of obesity compared with ob/ob only mice, as reflected by a strong reduction in body weight (21 +/- 4 vs. 44 +/- 7 g), total adipose tissue stores (15 +/- 3 vs. 25 +/- 3% body wt), and average adipocyte size (7,689 +/- 624 vs. 15,295 +/- 1,289 microm(2)). Although less pronounced, APOC1 overexpression also reduced body weight on a wild-type background, solely due to a reduction in adipose tissue. Furthermore, despite elevated plasma free fatty acid and triglyceride levels, APOC1 overexpression significantly improved insulin sensitivity in ob/ob mice, as demonstrated by a strong reduction in plasma glucose and insulin levels, as well as a better performance in the glucose tolerance test. In conclusion, a marked reduction in the uptake of fatty acids into adipocytes may underlie the protection from obesity and insulin resistance in transgenic mice overexpressing human APOC1.
Sigma-Aldrich
Pentadecanoic acid, 99%
Supelco
Pentadecanoic acid, analytical standard
Sigma-Aldrich
Pentadecanoic acid, ~99% (capillary GC)