MilliporeSigma
Human hypertension caused by mutations in WNK kinases.
Science (New York, N.Y.) (2001-08-11)
F H Wilson, S Disse-Nicodème, K A Choate, K Ishikawa, C Nelson-Williams, I Desitter, M Gunel, D V Milford, G W Lipkin, J M Achard, M P Feely, B Dussol, Y Berland, R J Unwin, H Mayan, D B Simon, Z Farfel, X Jeunemaitre, R P Lifton
ABSTRACT
Hypertension is a major public health problem of largely unknown cause. Here, we identify two genes causing pseudohypoaldosteronism type II, a Mendelian trait featuring hypertension, increased renal salt reabsorption, and impaired K+ and H+ excretion. Both genes encode members of the WNK family of serine-threonine kinases. Disease-causing mutations in WNK1 are large intronic deletions that increase WNK1 expression. The mutations in WNK4 are missense, which cluster in a short, highly conserved segment of the encoded protein. Both proteins localize to the distal nephron, a kidney segment involved in salt, K+, and pH homeostasis. WNK1 is cytoplasmic, whereas WNK4 localizes to tight junctions. The WNK kinases and their associated signaling pathway(s) may offer new targets for the development of antihypertensive drugs.
Product Description
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Furosemide
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®