MilliporeSigma
  • Stereoselective direct glycosylation with anomeric hydroxy sugars by activation with phthalic anhydride and trifluoromethanesulfonic anhydride involving glycosyl phthalate intermediates.
Stereoselective direct glycosylation with anomeric hydroxy sugars by activation with phthalic anhydride and trifluoromethanesulfonic anhydride involving glycosyl phthalate intermediates.
Journal of the American Chemical Society (2008-06-06)
Kwan Soo Kim, Dinanath Baburao Fulse, Ju Yuel Baek, Bo-Young Lee, Heung Bae Jeon
ABSTRACT
An efficient direct one-pot glycosylation method with anomeric hydroxy sugars as glycosyl donors employing phthalic anhydride and triflic anhydride as activating agents has been developed. Thus, highly stereoselective beta-mannopyranosylations were achieved by the reaction of 2,3-di-O-benzyl-4,6-O-benzylidene-D-mannopyranose (2) with phthalic anhydride in the presence of DBU at room temperature followed by sequential addition of DTBMP and Tf2O and glycosyl acceptors to the reaction mixture at -78 degrees C in one-pot. Stereoselective alpha-glucopyranosylations with 2,3-di-O-benzyl-4,6-O-benzylidene-D-glucopyranose (25) and other glycosylations with glucopyranoses and mannopyranoses having tetra-O-benzyl- and tetra-O-benzoyl protecting groups were also possible by utilizing the present one-pot glycosylation protocol. The possible mechanism for the beta-mannosylation with 2 was proposed based on the NMR study, in which alpha-mannosyl phthalate 55alpha and alpha-mannosyl triflate 59 were detected as intermediates. The versatility and efficiency of the present glycosylation methodology, especially those of the beta-mannopyranosylation protocol, were readily demonstrated by the efficient synthesis of protected beta-(1-->4)-D-mannotriose 62 and beta-(1-->4)-D-mannotetraose 67 with perfect beta-stereoselectivity.
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%