MilliporeSigma
  • A new method for determination of serum cholestanol by high-performance liquid chromatography with ultraviolet detection.
A new method for determination of serum cholestanol by high-performance liquid chromatography with ultraviolet detection.
Journal of chromatography. B, Biomedical sciences and applications (2000-07-20)
G Halperin, M Elisaf, E Leitersdorf, D Harats
ABSTRACT
We developed a method for the determination of serum 5alpha-cholestan-3beta-ol (cholestanol). The sterols were derivatized to the 4'-bromobenzenesulfonyl esters and heated in isopropanol. The cholesterol-4'-bromobenzenesulfonate was solvolyzed to cholesteryl isopropyl ether, but the derivatized cholestanol did not change and could be measured in a high-performance liquid chromatographic system equipped with a UV detector at 235 nm. On the other hand, the resulting cholesteryl isopropyl ether, having different absorbance and chromatographic mobility was not detected. This method was used for measuring cholestanol levels in patients with cerebrotendinous xanthomatosis (CTX), liver cirrhosis and serum from healthy control subjects. Reproducibility, linearity and recovery tests were done on 0.3 ml of serum samples containing >2 microg/ml cholestanol, using stigmastanol as an internal standard (I.S.). Determining cholestenol by this method can be used for diagnosis and follow-up of patients with CTX and various liver diseases.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Linoleic acid, analytical standard
SAFC
L-Aspartic acid
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Methanethiol, ≥98.0%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Tyramine, 98%, FG
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Creatine, anhydrous
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)