Merck
  • Solid-phase synthesis of imidazo[4,5-b]pyridin-2-ones and related urea derivatives by cyclative cleavage of a carbamate linkage.
Solid-phase synthesis of imidazo[4,5-b]pyridin-2-ones and related urea derivatives by cyclative cleavage of a carbamate linkage.
Journal of combinatorial chemistry (2002-07-09)
Monika Ermann, Nadja M Simkovsky, Stanley M Roberts, David M Parry, Andy D Baxter
ABSTRACT
A solid-phase synthesis of substituted cyclic urea derivatives as potential heterocyclic library scaffolds is described. 2-Amino-3-nitropyridine is attached to Wang resin via a carbamate linkage. Reduction of the nitro group was achieved with SnCl(2).2H(2)O. Reductive alkylation with a range of substituted benzaldehydes followed by cyclative cleavage afforded a small library of 3-substituted imidazo[4,5-b]pyridine-2-ones in 33-45% yield and 59-88% purity. Subsequently, this methodology was applied to the synthesis of 3-substituted imidazo[4,5-f]quinolin-2-ones.