MilliporeSigma
  • Application of ion-exchange cartridge clean-up in food analysis. I. Simultaneous determination of thiabendazole and imazalil in citrus fruit and banana using high-performance liquid chromatography with ultraviolet detection.
Application of ion-exchange cartridge clean-up in food analysis. I. Simultaneous determination of thiabendazole and imazalil in citrus fruit and banana using high-performance liquid chromatography with ultraviolet detection.
Journal of chromatography. A (1998-08-06)
Y Ito, Y Ikai, H Oka, J Hayakawa, T Kagami
ABSTRACT
A simple, rapid and reproducible analytical method for thiabendazole (TBZ) and imazalil (IMA) in citrus fruit and banana has been developed. The method involves the use of an ion-exchange cartridge for sample clean-up followed by ion-pair high-performance liquid chromatography with ultraviolet detection. The recoveries of TBZ and IMA from citrus fruits spiked at levels of 10 microgram/g and 5 microgram/g were in the range of 94-98% and 93-98% with coefficients of variation of 0.5-2.2% and 1.6-2.7%, respectively. The recoveries of TBZ and IMA from banana spiked at levels of 3 microgram/g and 2 microgram/g were 94% and 94% with coefficients of variation 1.1% and 4.9%, respectively. The detection limits for TBZ and IMA were 0.1 microgram/g in citrus fruit and 0.05 microgram in banana.
Sigma-Aldrich
Sodium 1-dodecanesulfonate, ReagentPlus®, ≥99%