MilliporeSigma
  • Gas chromatographic-mass spectrometric characterization of flavanones in citrus and grape juices.
Gas chromatographic-mass spectrometric characterization of flavanones in citrus and grape juices.
The Analyst (1992-07-01)
C S Creaser, M R Koupai-Abyazani, G R Stephenson
ABSTRACT
A method for the characterization of flavanones in fruit juices, involving solvent extraction, hydrolysis to the corresponding aglycones, trimethylsilylation and combined gas chromatography-mass spectrometry, is reported. The application of the method is demonstrated for the analysis of orange, lemon, grapefruit and grape juices.