MilliporeSigma
  • Determination of aloesin in plasma by high-performance liquid chromatography as fluorescent 9-anthroyl derivative.
Determination of aloesin in plasma by high-performance liquid chromatography as fluorescent 9-anthroyl derivative.
Archives of pharmacal research (1998-12-30)
K H Kim, J G Lee, J H Park, Y G Shin, S K Lee, T H Cho, S T Oh
ABSTRACT
A sensitive high-performance liquid chromatographic (HPLC) method for the determination of aloesin in plasma was developed. After solid-phase extraction from plasma and derivatization of aloesin and compound AD-1, which was prepared from aloesin as a internal standard, with 9-anthroylnitrile in the presence of quinuclidine, the derivatives were separated on a inertsil ODS-3 column using acetonitrile/methanol/water (3:1:6) as a mobile phase, and detected fluorimetrically at 460 nm with excitation at 360 nm. The detection limit of aloesin was 3.2 ng/ml in plasma (S/N = 3).
Sigma-Aldrich
9-Anthracenecarbonitrile, 97%
Pricing and availability is not currently available.