MilliporeSigma
  • Buccal cell DNA yield, quality, and collection costs: comparison of methods for large-scale studies.
Buccal cell DNA yield, quality, and collection costs: comparison of methods for large-scale studies.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2002-10-12)
Irena B King, Jessie Satia-Abouta, Mark D Thornquist, Jeannette Bigler, Ruth E Patterson, Alan R Kristal, Ann L Shattuck, John D Potter, Emily White, Jessie Satia Abouta
ABSTRACT
There is considerable interest in noninvasive and cost-effective methods for obtaining DNA in large-scale studies. In this randomized crossover study of 22 participants, we compared the DNA yield, quality, and associated costs of buccal cell DNA collected using cytobrushes (three brushes per collection) and swish (i.e., mouthwash) in self-administered procedures. There was a nonstatistically significant higher yield from the mouthwash compared with cytobrush collections (15.8 microg versus 12.0 microg, respectively; P = 0.53). PCR reactions that required short (0.3 kb) or intermediate (1.1 kb) DNA fragments were 100% successful for DNA from brush and mouthwash, whereas PCRs for reactions that required long fragments (7.8 kb) failed for all of the participants from cytobrush DNA and were 81% successful for DNA from the mouthwash source. The brush collections provided sufficient DNA for an estimated 150-225 PCR reactions requiring short and intermediate DNA fragments. The estimated per person costs for buccal brush DNA collections in large studies were less then half (8.50 dollars) those for the mouthwash method (18 dollars). In addition, we tested whether cytobrush instructions to rub cheeks before collection or collect cells only in the morning increased DNA yield and whether repeat brushings of the same cheek reduced DNA yield. These variations resulted in no significant differences in DNA yields. We conclude that the collection of DNA with cytobrushes using simple instructions is cost effective in large-scale studies, and yields sufficient quantity and quality of DNA for genotyping.
Sigma-Aldrich
Deoxyribonucleic acid, low molecular weight from salmon sperm