MilliporeSigma
  • Multicenter, randomized, double-blind comparison of erythromycin estolate versus amoxicillin for the treatment of acute otitis media in children. AOM Study Group.
Multicenter, randomized, double-blind comparison of erythromycin estolate versus amoxicillin for the treatment of acute otitis media in children. AOM Study Group.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (1998-10-09)
H Scholz, R Noack
ABSTRACT
Erythromycin is frequently prescribed in Germany for acute otitis media, but well-designed clinical trials under present epidemiological conditions are lacking. Therefore, a double-blind, randomized, multicenter trial was performed to compare the clinical efficacy and safety of erythromycin estolate versus amoxicillin in children with acute otitis media and to identify the risk factors associated with clinical failure. Investigators from 19 centers throughout Germany recruited 302 children with clinical, otoscopic, and tympanometric evidence of acute otitis media. In a double-blind fashion, patients were allocated randomly to a 10-day course of erythromycin estolate at 40 mg/kg/day in two divided doses or amoxicillin at 50 mg/kg/day in two divided doses. Clinical examinations, otoscopy, and tympanometry were performed at baseline, day 3-5, day 9-11, and at 5 weeks. Clinical outcome was assessed on day 9-11. Two-hundred eighty children were evaluable for efficacy (erythromycin group, 141; amoxicillin group, 139). Both groups were comparable with respect to demographic data and severity of disease at entry. Treatment was successful in 94% of the erythromycin-treated patients and in 96% of the amoxicillin-treated patients. Clinical outcome was statistically equivalent between groups within a range of 7 percentage points. Clinical recurrence was seen in eight erythromycin-treated children (5.7%) and in seven amoxicillin-treated children (5.0%) (P=0.81). Patients with bilateral disease at entry were at higher risk of unfavourable outcome, whereas age and presence/absence of otorrhea at entry were not associated with outcome. Treatment-related adverse events were recorded in eight (5.3%) of 151 erythromycin-treated patients and in 11 (7.3%) of 151 amoxicillin-treated patients. In this study in an outpatient setting in Germany, erythromycin estolate was as safe and effective as amoxicillin in the treatment of acute otitis media. Both drugs can be administered in a convenient twice-daily dosage schedule.
Product Description
Erythromycin estolate, European Pharmacopoeia (EP) Reference Standard