MilliporeSigma
  • Inhibition by platelet-activating factor of Src- and hepatocyte growth factor-dependent invasiveness of intestinal and kidney epithelial cells. Phosphatidylinositol 3'-kinase is a critical mediator of tumor invasion.
Inhibition by platelet-activating factor of Src- and hepatocyte growth factor-dependent invasiveness of intestinal and kidney epithelial cells. Phosphatidylinositol 3'-kinase is a critical mediator of tumor invasion.
The Journal of biological chemistry (1998-06-11)
L Kotelevets, V Noë, E Bruyneel, E Myssiakine, E Chastre, M Mareel, C Gespach
ABSTRACT
This study was designed to characterize platelet-activating factor receptor (PAF-R) expression and function in normal and cancerous human colonic epithelial cells. PAF-R gene transcripts were analyzed by reverse transcription-polymerase chain reaction and Southern blot, using three sets of primers corresponding either to the coding region of the human PAF-R sequence (polymerase chain reaction product: 682 base pairs (bp)) or to the leukocyte- and tissue-type transcripts of 166 and 252 bp, respectively. An elongated splice variant was identified in the 5'-untranslated region of the tissue-type PAF-R transcript (334 bp) in colonic epithelial crypts and tumors. In human colonic PCmsrc cells transformed by c-src oncogene, the hepatocyte growth factor (HGF)-dependent invasiveness of collagen gels was abolished by 0.1 microM PAF and restored by the PAF-R antagonists WEB2086 and SR27417. PAF blocked HGF-induced tyrosine phosphorylation of p125 focal adhesion kinase. The phosphatidylinositol 3'-kinase (PI3'-K) inhibitors wortmannin and LY294002 totally blocked the HGF-induced invasion. Similar effects were observed in ts-srcMDCK kidney epithelial cells transformed by a v-Src temperature-sensitive mutant: (i) PAF and wortmannin exerted additive inhibitory effects on Src-induced invasion and (ii) activated and dominant negative forms of p110alpha PI3'-K, respectively, amplified and abrogated the Src- and HGF-dependent invasiveness of parental and ts-srcMDCK cells. We also provided the first evidence for the contribution of rapamycin-insensitive, pertussis toxin-dependent G-protein pathways to the integration of the signals emerging from activated Met and PAF receptors. These results indicate that PI3'-K is a critical transducer of invasiveness and strongly suggest that PAF exerts a negative control on invasion by inhibiting this signaling pathway. A possible beneficial role of PAF analogs on tumor invasion is therefore proposed.
Sigma-Aldrich
SR 27417, ≥98% (HPLC)