MilliporeSigma
  • Affinity labelling of frog brain opioid receptors by dynorphin(1-10) chloromethyl ketone.
Affinity labelling of frog brain opioid receptors by dynorphin(1-10) chloromethyl ketone.
Neuropeptides (1997-02-01)
S Benyhe, A Ketevan, J Simon, J Hepp, K Medzihradszky, A Borsodi
ABSTRACT
It has been previously found that chloromethyl ketone derivatives of enkephalins bind irreversibly to the opioid receptors in vitro. Recently a novel affinity reagent, Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Ile-Arg-Pro-Gly chloromethyl ketone (Dynorphin(1-10)-Gly11 chloromethyl ketone, DynCMK) was synthesized, and its binding characteristics to frog (Rana esculenta) brain membranes were evaluated. In competition experiments, the product shows a relatively high affinity for the kappa-opioid binding sites labelled by [3H]ethylketocyclazocine (Ki is approximately equal to 200 nM), whereas its binding to the 1 ([3H]dihydromorphine) and to the delta sites ([3H]D-Ala2-Leu5]enkephalin) is weaker. Preincubation of the frog brain membranes with DynCMK at micromolar concentrations results in a washing-resistant and dose-dependent inhibition of the [3H]ethylketocyclazocine binding sites. Saturation binding analysis of the membranes preincubated with 50 microM DynCMK reveals a significant decrease in the number of specific binding sites for [3H]ethylketocyclazocine compared to the control values. The kappa-preferring binding properties of the compound suggest that it could serve as an affinity label for the kappa-type of opioid receptors.