MilliporeSigma
  • Gamma-mangostin, a novel type of 5-hydroxytryptamine 2A receptor antagonist.
Gamma-mangostin, a novel type of 5-hydroxytryptamine 2A receptor antagonist.
Naunyn-Schmiedeberg's archives of pharmacology (1998-02-12)
N Chairungsrilerd, K I Furukawa, T Ohta, S Nozoe, Y Ohizumi
ABSTRACT
Gamma-mangostin, purified from the fruit hull of the medicinal plant Garcinia mangostana caused a parallel rightwards shift of the concentration/response curve for the contraction elicited by 5-hydroxytryptamine (5-HT) in the rabbit aorta (pA2 = 8.2) without affecting the contractile responses to KCl, phenylephrine (alpha1) or histamine (H1). The perfusion pressure response of rat coronary artery to 5-HT (5-HT2A) was reduced concentration dependently by gamma-mangostin (IC50 = 0.32 microM). 5-HT amplified, ADP-induced aggregation of rabbit platelets (5-HT2A) was inhibited by gamma-mangostin (IC50 = 0.29 microM), whereas that induced by thrombin was not affected, nor did gamma-mangostin affect 5-HT-induced contraction of the guinea-pig ileum (5-HT3)in the presence of 5-HT1, 5-HT2 and 5-HT4 receptor antagonists. Furthermore, 5-HT-induced contraction of the rat fundus (5-HT2B) and 5-HT-induced relaxation of the rabbit aorta in the presence of ketanserin (5-HT1) and carbachol-induced contraction of the guinea-pig ileum (muscarinic M3) were not affected by gamma-mangostin (5 microM). Gamma-mangostin inhibited [3H]spiperone binding to cultured rat aortic myocytes (IC50 = 3.5 nM). The Kd for [3H]spiperone binding was increased by gamma-mangostin (3 nM) from 11.7 to 27.4 nM without affecting Bmax. These results suggest that gamma-mangostin is a novel competitive antagonist, free from a nitrogen atom, for the 5-HT2A receptors in vascular smooth muscles and platelets.
Sigma-Aldrich
gamma-Mangostin, ≥98% (HPLC)