MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Psychosis during pregnancy: treatment considerations.
Annals of clinical psychiatry : official journal of the American Academy of Clinical Psychiatrists (1997-10-27)
H B Pinkofsky
ABSTRACT
The onset of psychosis during pregnancy presents several difficult management decisions and a careful risk-benefit analysis is required. Withholding antipsychotic treatment may produce more risks than benefits. Studies on neuroleptic teratogenicity are contradictory. Most of the commonly used neuroleptics exhibit a pregnancy risk of category C. Neuroleptic use during pregnancy may be associated with adverse effects in the pre- and postnatal period. These concerns include compromising uterine blood flow, post-partum neonatal sedation, and extrapyramidal signs expressed in the neonate. Each neuroleptic exhibits a unique pharmacokinetic profile. The antipsychotic properties and side effects considered most significant include sedation, half-life, hypotension, and apparent hydrophilicity. In this case study a decision to select molindone was based on these parameters.