MilliporeSigma
  • Differential involvement of disulfide bridges on the folding of a scorpion toxin.
Differential involvement of disulfide bridges on the folding of a scorpion toxin.
The journal of peptide research : official journal of the American Peptide Society (1997-07-01)
V Calabro, J M Sabatier, E Blanc, C Lecomte, V Van Rietschoten, H Darbon
ABSTRACT
Leiurotoxin I is a neurotoxin, blocker of Ca(2+)-activated apamin-sensitive K+ channel, purified from the venom of the scorpion Leiurus quinquestriatus hebraeus. It is a 31-residue polypeptide reticulated by three disulfide bridges, i.e. Cys3-Cys21, Cys8-Cys26 and Cys12-Cys28. To investigate the role of these disulfide bridges in the folding of this toxin, analogs lacking one disulfide bridge were synthesized. The structures of two analogs in which two half-cystines were placed by alpha-aminobutyrate residues to suppress one disulfide bridge, were analyzed by 1H NMR. The NMR studies reveal a three-dimensional structure identical with the native toxin for the analog lacking disulfide bridge Cys3-Cys21 and a loss of organized structure for another analog lacking disulfide bridge Cys12-Cys28. These analogs are, respectively, fully active and weakly active (2% of the residual activity) when tested in vitro for their ability to interact with their receptor channel and in vivo for their neurotoxic activity in mice. This suggest that disulfide bridge Cys12-Cys28 is essential for the folding process. In contrast, the lack of disulfide bridge Cys3-Cys21 does not affect the folding and the maintenance of bioactive conformation of Leiurotoxin I.