MilliporeSigma
  • The photodynamic effect of Victoria blue BO on peripheral blood mononuclear and leukemic cells.
The photodynamic effect of Victoria blue BO on peripheral blood mononuclear and leukemic cells.
Photochemistry and photobiology (1997-05-01)
M Fiedorowicz, A Pituch-Noworolska, M Zembala
ABSTRACT
The photodynamic effect of Victoria blue BO (VB-BO) and photoirradiation on peripheral blood mononuclear cells was studied. The cells were preincubated with VB-BO followed by photoirradiation and overnight culture. The highest percentage of dead cells (propidium iodide assay in flow cytometry) was seen in the monocyte population. The lymphocytes showed a lower sensitivity to VB-BO photodynamic action than the monocytes (12% vs 80% of PI-positive cells). The effect of VB-BO and phototreatment on lymphocyte function was studied using a mitogen-induced proliferation assay. A decrease of mitogen response was observed. The VB-BO and photoirradiation were also used on leukemic cells. The leukemic cells from acute myeloid leukemia and B precursors leukemia were sensitive to VB-BO photodynamic action. The high VB-BO sensitivity of monocytes and leukemic cells (myeloid and lymphoid B derived) suggests possible application of VB-BO for selective depletion of monocytes or sensitive leukemic cells.
Sigma-Aldrich
Victoria Pure Blue BO, Dye content 90 %