MilliporeSigma
  • Modulation of neurotransmitter release by opioid mu- and kappa-receptors from adrenergic terminals in the myenteric plexus of the guinea-pig colon: effect of alpha 2-autoreceptor blockade.
Modulation of neurotransmitter release by opioid mu- and kappa-receptors from adrenergic terminals in the myenteric plexus of the guinea-pig colon: effect of alpha 2-autoreceptor blockade.
Neuroscience letters (1997-01-31)
M Cosentino, F Marino, R Bombelli, M Ferrari, E Rasini, C Giaroni, S Lecchini, G Frigo
ABSTRACT
We have studied the effect of [D-Ala2,N-Me-Phe4,Gly-ol5]-enkephalin (DAMGO, opioid mu-receptor agonist) and ICI-204,448 (kappa-receptor agonist) on endogenous noradrenaline release in the guinea-pig isolated distal colon. DAMGO enhances noradrenaline over-flow and this effect is antagonized by naloxone (pIC50 = 10.27) and nor-binaltorphimine (pIC50 = 7.97), and concentration-dependently turned into inhibition by yohimbine. ICI-204,448 inhibits noradrenaline overflow and is antagonized by naloxone (pIC50 = 9.38) and nor-binaltorphimine (pIC50 = 10.48), but is not affected by yohimbine. Evidence is thus given that mu- and kappa-opioid receptors modulate noradrenaline release in the guinea-pig colon. Modifications by yohimbine of the effect of DAMGO indicate the existence of a functional relationship between mu-receptors and alpha(2)-autoreceptors in this model.