MilliporeSigma
  • An anti-cancer derivative of butyric acid (pivalyloxmethyl buterate) and daunorubicin cooperatively prolong survival of mice inoculated with monocytic leukaemia cells.
An anti-cancer derivative of butyric acid (pivalyloxmethyl buterate) and daunorubicin cooperatively prolong survival of mice inoculated with monocytic leukaemia cells.
British journal of cancer (1997-01-01)
T Kasukabe, A Rephaeli, Y Honma
ABSTRACT
A derivative of butyric acid, pivalyloxymethyl butyrate (AN-9), inhibited the proliferation and induced apoptosis of mouse monocytic leukaemia Mm-A cells, although sodium butyrate, but not AN-9, induced differentiation of the cells. AN-9 and DNA-specific antineoplastic agents synergistically inhibited the growth of Mm-A cells, and the simultaneous treatment was required to evoke the maximum growth-inhibitory effect. On the other hand, there was no synergy between butyrate and the drugs, or AN-9 and anti-metabolic agents in inhibiting the growth of the cells, suggesting that the synergistic effect is specific to AN-9 and DNA-reacting agents. AN-9 as a single agent prolonged the survival of mice inoculated with Mm-A cells in a dose-dependent manner. Moreover, administration of AN-9 plus daunorubicin (DNR) markedly prolonged their survival. These results suggest that combination with AN-9 and DNR entails an obvious therapeutic potential.