MilliporeSigma
  • Effect of S-ethylisothiourea, a putative inhibitor of inducible nitric oxide synthase, on mouse skin vascular permeability.
Effect of S-ethylisothiourea, a putative inhibitor of inducible nitric oxide synthase, on mouse skin vascular permeability.
Japanese journal of pharmacology (1996-03-01)
T Muraki, E Fujii, M Okada, H Horikawa, K Irie, K Ohba
ABSTRACT
By dye leakage in mouse skin, we evaluated the inhibition of proinflammatory stimuli-induced plasma extravasation by a putative inhibitor of inducible nitric oxide synthase, S-ethylisothiourea. A low dose of S-ethylisothiourea (5 micrograms/kg) mimicked aminoguanidine in inhibiting the plasma extravasation elicited by lipopolysaccharide but not by 5-hydroxytryptamine or platelet-activating factor. A higher dose of S-ethylisothiourea (10 micrograms/kg) inhibited the plasma extravasation induced by 5-hydroxytryptamine slightly; however, it increased the basal dye leakage. Thus, S-ethylisothiourea may be used as a relatively specific inhibitor for inducible nitric oxide synthase in vivo.