MilliporeSigma
  • Speciation of arsenic oxides using laser desorption/ionization time-of-flight mass spectrometry.
Speciation of arsenic oxides using laser desorption/ionization time-of-flight mass spectrometry.
Analytical chemistry (1996-11-15)
T M Allen, D Z Bezabeh, C H Smith, E M McCauley, A D Jones, D P Chang, I M Kennedy, P B Kelly
ABSTRACT
Positive and negative ion mass spectra of arsenic trioxide (As2O3) and arsenic pentaoxide (As2O5) have been obtained by single-step laser desorption/ionization time-of-flight mass spectrometry. Pulsed UV radiation at 266 nm was used for the simultaneous desorption and ionization of the solid sample. High-mass cluster ions that are unique to the oxidation state of each oxide sample appear in the negative ion mass spectra. The As2O3 produces As3O5-, while the As2O5 yields As3O8-. The formation of unique negative cluster ions presents the capability for arsenic oxidation state speciation by laser desorption/ionization mass spectrometry. The ability of time-of-flight mass spectrometry to examine the relative amounts of each arsenic oxide present in a series of mixtures is discussed. Application of our speciation technique to a model incinerator sample is demonstrated.