MilliporeSigma
  • Antiprogestins inhibit the binding of opioids to mu-opioid receptors in nervous membrane preparations.
Antiprogestins inhibit the binding of opioids to mu-opioid receptors in nervous membrane preparations.
European journal of pharmacology (1996-04-22)
R Maggi, F Pimpinelli, L A Casulari, F Piva, L Martini
ABSTRACT
The present study showed that the glucocorticoid/progesterone antagonists, 17 beta-hydroxy-1 1 beta-(4-dimethylamino-phenyl-1)-17-(prop-1-ynyl)estra-4,9-dien+ ++-3-one (RU486) and 17 beta-hydroxy-11 beta-(4-dimethylamino-phenyl-1)-17-(propan-3-ol)estra-4,9-dien-3-o ne (ZK 98299), inhibit the binding of labeled dihydromorphine to mu-opioid receptors present on membrane preparations derived from rat and mouse brain, as well as from human neuroblastoma cells. The inhibitory effect of RU486 was dose-dependent and linked to a decrease of the affinity of labeled dihydromorphine to the mu-opioid receptors. Kinetic experiments have shown that RU486 induces a decrease of the association rate constant (k + 1) of dihydromorphine. RU486 also proved able to dissociate the dihydromorphine-mu-opioid receptor complex, although at a rate slower than that exhibited by unlabeled dihydromorphine. Finally, the addition of NaCl (100 mM) to the incubation buffer induced a 50% decrease of the inhibitory effect of RU486. A 6-day treatment of neuroblastoma cells with RU486 eliminated the inhibitory effect morphine exerts on the intracellular accumulation of cyclic AMP induced by prostaglandin E1. These results indicate that RU-486 may interact with brain mu-opioid receptors in vitro, by decreasing the affinity of opioid ligands.