MilliporeSigma
  • Comparative cytoprotection of cultured corneal endothelial cells by water-soluble antioxidants against free-radical damage.
Comparative cytoprotection of cultured corneal endothelial cells by water-soluble antioxidants against free-radical damage.
Cornea (1995-09-01)
L H Zeng, D S Rootman, K P Fung, T W Wu
ABSTRACT
We reported previously that purpurogallin (PPG) markedly protects the cultured rabbit corneal endothelial cells (RCEC) against oxyradical damage generated with hypoxanthine (HX) and xanthine oxidase (XO)(1). In this study, we further compared the cytoprotective activities of PPG versus Trolox (TX, alpha-tocopherol, a water-soluble analogue of vitamin E) and ascorbate (Asc) in confluent cultured RCEC with phase contrast microscopy and confirmed by transmission electron microscopy. PPG prolonged survival of the oxyradical damaged cells longer than those without PPG present (18.6 +/- 1.4 min at 1.0 mM and 11.2 +/- 1.0 at 0.25 mM respectively vs. 7.3 +/- 0.8 min in control). At levels equimolar to PPG, TX, and Asc were less effective in delaying cell necrosis caused by HX and XO (p < 0.01). When exposed to superoxide radicals generated by menadione, RCEC necrosed at 29.8 +/- 1.5 min compared to PPG 47.2 +/- 1.0 min at 1.0 mM and 38.9 +/- 1.0 min at 0.25 mM. This was significantly different from TX and Asc at corresponding concentrations (p < 0.01). PPG scavenges not only HX-XO-generated oxyradicals, but also nonenzymatically produced superoxide radicals, more actively than two well known antioxidants--TX and Asc.