MilliporeSigma
  • Protection against acute MPTP-induced dopamine depletion in mice by adenosine A1 agonist.
Protection against acute MPTP-induced dopamine depletion in mice by adenosine A1 agonist.
Journal of neurochemistry (1993-02-01)
Y S Lau, M M Mouradian
ABSTRACT
The effects of the adenosine A1 agonist N6-cyclohexyladenosine (CHA) on MPTP-induced dopamine (DA) depletion in the striatum of C57BL/6 mice were studied. Twenty hours after a single injection of MPTP (30 mg/kg, s.c.), the toxin caused 62% depletion of striatal DA. CHA (0.2-3 mg/kg, s.c.), when given together with MPTP, prevented the toxin-induced DA depletion in a dose-dependent manner. This protective action was apparently mediated by the A1 receptors, because this effect was selectively antagonized by pretreating the animals with the A1 antagonist 8-cyclopentyl-1,3-dipropylxanthine (25 mg/kg, i.p.) but not with the A2 antagonist 1,3-dipropyl-7-methylxanthine (25 mg/kg, i.p.). When CHA (3 mg/kg) was injected 5 h after MPTP administration, at which point striatal DA levels were already reduced significantly, a rapid and complete recovery of the striatal DA levels occurred. These neurochemical data suggest that the A1 agonist CHA is potentially useful as a neuroprotective agent against MPTP-induced toxicity.